Mercury standard solution, for ICP, in HNO₃, CRM, VHG
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VHG-PHGN-100 VHG-PHGN-500 VHG-THGN-100 VHG-THGN-500 VHG-LHGN-100 VHG-LHGN-250
PROOVHG-PHGN-100EA 0 EUR
PROOVHG-PHGN-100 PROOVHG-PHGN-500 PROOVHG-THGN-100 PROOVHG-THGN-500 PROOVHG-LHGN-100 PROOVHG-LHGN-250
Mercury standard solution, for ICP, in HNO₃, CRM, VHG
Standaarden Kationische standaarden Kwik standaarden
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certificaten: Traceable to NIST, SRM.