Padlock control centres
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
800118 800119
BRDY800118EA 343.81 EUR
BRDY800118 BRDY800119
Padlock control centres
Veiligheidssloten
These padlock control centres provide safe storage and controlled access to padlocks.
  • Made from powder-coated steel
  • Each nickel coated metal hook can hold two locks
  • Two keys are provided for cabinet lock
Bestelinformatie: Components not included. Padlocks need to be ordered separately.
Order Now
Specificaties
Environmentally Preferable