Holmium standard solution, for ICP, in 5% HNO₃, VHG
Leverancier: LGC Standards PROMOCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VHG-PHON-100
VHG-PHON-500
VHG-THON-100
VHG-THON-500
PROOVHG-PHON-100EA
0
EUR
PROOVHG-PHON-100
PROOVHG-PHON-500
PROOVHG-THON-100
PROOVHG-THON-500
Holmium standard solution, for ICP, in 5% HNO₃, VHG
Standaarden
Kationische standaarden
Holmium standaarden
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certificaten: Traceable to NIST, SRM.