Indium standard solution, for ICP, in 5% HNO₃, VHG
Leverancier: LGC Standards PROMOCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VHG-PINN-100
VHG-PINN-500
VHG-TINN-100
VHG-TINN-500
PROOVHG-PINN-100EA
0
EUR
PROOVHG-PINN-100
PROOVHG-PINN-500
PROOVHG-TINN-100
PROOVHG-TINN-500
Indium standard solution, for ICP, in 5% HNO₃, VHG
Standaarden
Kationische standaarden
Indium standaarden
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certificaten: Traceable to NIST, SRM.