Molybdenum standard solution, for ICP, in NH₄OH, VHG
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VHG-PMOZ-100 VHG-PMOZ-500 VHG-TMOZ-100 VHG-TMOZ-500
PROOVHG-PMOZ-100EA 0 EUR
PROOVHG-PMOZ-100 PROOVHG-PMOZ-500 PROOVHG-TMOZ-100 PROOVHG-TMOZ-500
Molybdenum standard solution, for ICP, in NH₄OH, VHG
Standaarden Kationische standaarden Molybdeen standaarden
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certificaten: Traceable to NIST, SRM.