Nickel standard solution, for ICP, in 5% HNO₃, VHG
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VHG-PNIN-100 VHG-PNIN-500 VHG-PNIN-50 VHG-TNIN-100 VHG-TNIN-500
PROOVHG-PNIN-100EA 0 EUR
PROOVHG-PNIN-100 PROOVHG-PNIN-500 PROOVHG-PNIN-50 PROOVHG-TNIN-100 PROOVHG-TNIN-500
Nickel standard solution, for ICP, in 5% HNO₃, VHG
Standaarden Kationische standaarden Nikkel standaarden
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certificaten: Traceable to NIST, SRM.