Osmium standard solution, for ICP, in 20% HCl, VHG
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VHG-POSH-100 VHG-POSH-50 VHG-POSH-500
PROOVHG-POSH-100EA 0 EUR
PROOVHG-POSH-100 PROOVHG-POSH-50 PROOVHG-POSH-500
Osmium standard solution, for ICP, in 20% HCl, VHG
Standaarden Kationische standaarden Osmium standaarden
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certificaten: Traceable to NIST, SRM.