Lead standard solution, for ICP, in HNO₃, VHG
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VHG-PPBN-100 VHG-PPBN-500 VHG-LPBN-100 VHG-LPBN-500 VHG-PPBN-50 VHG-TPBN-100 VHG-TPBN-500
PROOVHG-PPBN-100EA 0 EUR
PROOVHG-PPBN-100 PROOVHG-PPBN-500 PROOVHG-LPBN-100 PROOVHG-LPBN-500 PROOVHG-PPBN-50 PROOVHG-TPBN-100 PROOVHG-TPBN-500
Lead standard solution, for ICP, in HNO₃, VHG
Standaarden Kationische standaarden Lood standaarden
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certificaten: Traceable to NIST, SRM.
Order Now
Specificaties
Environmentally Preferable