Bottles, wide neck, round, HDPE, with sealing cap
Leverancier: LAMAPLAST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
435 434 433
215-4161EA 387 EUR
215-4161 215-4160 215-4159
Bottles, wide neck, round, HDPE, with sealing cap
Flessen Kunststof flessen
HDPE, white with self sealing white PP screw cap.
  • For food use
  • Temperature range: -40 to +80 °C
  • Sterilisable by radiation
  • Not autoclavable
The bottles are suitable for food contact. They meets relevant requirements of the following directives or regulations: 1935/2004/CE as amended, UE 10/2011 as amended.