Ammonium ion chromatography standards, VHG
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VHGINH3-1K-100 VHGINH3-1K-500
PROOVHGINH3-1K-100EA 0 EUR
PROOVHGINH3-1K-100 PROOVHGINH3-1K-500
Ammonium ion chromatography standards, VHG
Standaarden Kationische standaarden Ammonium standaarden
These standard solutions are intended for use as a calibration or reference standard for ion chromatography (IC) or techniques using other modes of aqueous ion detection.
Certificaten: Traceable to NIST and CRM.