Barium standard solution, for ICP, in 5% HNO₃, VHG
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VHG-PBAN-100 VHG-TBAN-100 VHG-PBAN-50 VHG-PBAN-500 VHG-TBAN-500
PROOVHG-PBAN-100EA 0 EUR
PROOVHG-PBAN-100 PROOVHG-TBAN-100 PROOVHG-PBAN-50 PROOVHG-PBAN-500 PROOVHG-TBAN-500
Barium standard solution, for ICP, in 5% HNO₃, VHG
Standaarden Kationische standaarden Barium standaarden
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certificaten: Traceable to NIST, SRM.