Water standards for oils, Karl Fischer titration, CRM, VHG
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VHGKF0.05P100 VHGKF-0.5P-100 VHG-KF-1.0P100 VHG-KF-0.1P100
PROOVHGKF0.05P100EA 0 EUR
PROOVHGKF0.05P100 PROOVHGKF-0.5P-100 PROOVHG-KF-1.0P100 PROOVHG-KF-0.1P100
Water standards for oils, Karl Fischer titration, CRM, VHG
Standaarden Standaarden voor waterbepaling (door de Karl Fischer methode)
This solution is intended for use as a certified reference material in the determination of water in motor oil by Karl Fischer titration.
Certificaten: Traceable to NIST and SRM.
Order Now
Specificaties
Environmentally Preferable