Oils for vacuum pumps, DuoSeal®
Leverancier: Welch by Gardner Denver
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1407K-11 1407K-15
ILMV1407K-11EA 34.1 EUR
ILMV1407K-11 ILMV1407K-15
Oils for vacuum pumps, DuoSeal®
Smeermiddelen
A specially fractionated oil for DuoSeal® belt-driven pumps is designed to ensure the highest vacuum performance. The oil is tested to high vacuum levels to meet rigid requirements for vapor pressure, vacuum level stability, and viscosity. DuoSeal® oil is famous for its quality and consistency.