Doekjesrollen, WYPALL* L30 ULTRA+
Leverancier: KIMBERLY CLARK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7331 7425 7426
115-2108EA 153 EUR
115-2108 115-2217 115-2218
Doekjesrollen, WYPALL* L30 ULTRA+
Schoonmaakdoekjes Doekjes en handdoeken voor eenmalig gebruik Doekjes voor algemeen gebruik
These single use wipes made from three ply AIRFLEX* material are ideal for superior cleaning and heavy duty wiping tasks such as soaking up industrial spills, cleaning parts and components.
  • Strongest, thickest and most absorbent WYPALL L wipe
  • Stronger than conventional three ply wipes
  • Available as a white or blue perforated large roll
  • Can be used with a dispensing system to help control use and reduce wastage
For use with dispensers 115-0045, 115-1533 and 115-2038.