Human cord blood, unprocessed
Leverancier: Charles River Laboratories Cell Solutions
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CB008F-0 CB008F-1
CB008F-0EA 0 EUR
CB008F-0 CB008F-1
Human cord blood, unprocessed
Bloed
Cord blood is known to be a rich source of hematopoietic stem cells, which can be used to research hematopoietic and genetic disorders.
  • Test negative for HIV, HTLVI/II, HCV, HBV, WNV, syphilis, CMV, NAT, NAT WNV and Chagas
Human cord blood is collected aseptically at the time of delivery from healthy pregnant women. The donor is required to review and sign an approved procedure-specific consent form prior to the collection.
Storage: Use immediately upon receipt.
Opgelet: Intended for research use only and not for direct therapeutic or diagnosticMore Product Information
Order Now
Specificaties
Environmentally Preferable