Giemsa's stain for use in preparing solutions of leishman's, jenner's, giemsa's and other stains
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Azure mixture sicc. Giemsa stain, Azur-Eosin-Methylene blue acc. Giemsa, Giemsa's azur-eosin-methylene blue
B21172.09 B21172.18
B21172.09EA 28.4 EUR
B21172.09 B21172.18
Giemsa's stain for use in preparing solutions of leishman's, jenner's, giemsa's and other stains
Giemsa's stain
Giemsa stain is a laboratory reagent and stain for microscopy used in cytogenetics and histopathologic detection of malaria and other parasites such as spirochete and protozoan blood parasites.
The stain can be incorporated as a differential staining between bacterial and human cells.
It can also be used to visualize chromosomes or to identify mast cells.
Smeltpunt: ∼ 300 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00081642
CAS nummer: 51811-82-6
EINECS: 257-438-2