Cryoboxes, Cardboard
Leverancier: RATIOLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5131506 5160028 5120128 5160029
RATI5131506EA 35.3 EUR
RATI5131506 RATI5160028 RATI5120128 RATI5160029
Cryoboxes, Cardboard
Dozen Dozen voor cryobuisjes
Cryoboxes, cryo resistant cardboard.
  • Temperature resistant down to -80 °C
  • Wwater-repellent finish
For storage of cryovials at low temperatures