Ion chromatography standard, lithium, VHG
Leverancier: LGC Standards PROMOCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VHG-ILI100-500
VHG-ILIW1K-100
PROOVHG-ILI100-500EA
0
EUR
PROOVHG-ILI100-500
PROOVHG-ILIW1K-100
Ion chromatography standard, lithium, VHG
Standaarden
Kationische standaarden
Lithium standaarden
This solution is intended for use as a certified reference material and calibration standard, for ion chromatography (IC) and techniques using other modes of aqueous ion detection.
Certificaten: Traceable to NIST and SRM.