Phosphate ion chromatography standard (PO₄⁻³ as P), VHG
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VHG-IPO4P-100 VHG-IPO4P-500
PROOVHG-IPO4P-100EA 0 EUR
PROOVHG-IPO4P-100 PROOVHG-IPO4P-500
Phosphate ion chromatography standard (PO₄⁻³ as P), VHG
Standaarden Anionische standaarden Fosfaat(P) standaarden
This solution is intended for use as a certified reference material (CRM) and calibration standard, for ion chromatography (IC) and techniques using other modes of aqueous ion detection.
Certificaten: Traceable to NIST and SRM.
Order Now
Specificaties
Environmentally Preferable