Volumetric flasks for crude oil analysis, glass, clear
Leverancier: NEUBERT VOLUME GLASSWAERE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0378-70-100190 0378-70-250 0378-70-087 0378-70-088 0378-70-089
NEUB0378-70-100190EA 206 EUR
NEUB0378-70-100190 NEUB0378-70-250 NEUB0378-70-087 NEUB0378-70-088 NEUB0378-70-089
Volumetric flasks for crude oil analysis, glass, clear
Kolven Maatkolven
Graduated trapezoidal shaped flasks of borosilicate glass 3.3 with glass stoppers.
Used for the determination of the acid-separable fraction of water-miscible cooling lubricants.
Certificaten: DIN 51368