Dissolved oxygen meter, bench, Eutech DO 700
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Eutech DO 700
664-0233EA 1197 EUR
664-0233
Dissolved oxygen meter, bench, Eutech DO 700
Elektrochemische meters Zuurstofmeters
This economical and user-friendly DO meter is ideal for routine applications in laboratories, productions plants and schools.
  • Dissolved oxygen measurements in ppm, mg/L or % saturation
  • Large, easy-to-read display
  • Ready indicator informs users when readings are stable
  • Accurate readings in varying conditions with temperature, salinity and barometric pressure compensation
  • Non volatile memory holds up to 100 data points
  • Integral electrode holder
Leveringsinformatie: Supplied with dissolved oxygen/ATC probe (DO6HANDY), integral electrode holder and 100/240 VAC adapter.