Sulphate ion chromatography standard, VHG
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VHG-ISO41K-100 VHG-ISO41K-500 VHG-I1PSO4-100
PROOVHG-ISO41K-100EA 0 EUR
PROOVHG-ISO41K-100 PROOVHG-ISO41K-500 PROOVHG-I1PSO4-100
Sulphate ion chromatography standard, VHG
Standaarden Anionische standaarden Sulfaat standaarden
This standard solution is intended for use as a certified reference material (CRM) and calibration standard, for ion chromatography (IC) and techniques using other modes of aqueous ion detection.
Certificaten: Traceable to NIST and SRM.