Autosampler caps and septa
Leverancier: PerkinElmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
N9302685 N9302686 N9306015 N9302684 N9306052 N9306203 N9306205 N9306228 N9306229 N6356473 N9303419 N9303449 N9306360 N9306362 N9306364
PERCN9302685EA 60.2 EUR
PERCN9302685 PERCN9302686 PERCN9306015 PERCN9302684 PERCN9306052 PERCN9306203 PERCN9306205 PERCN9306228 PERCN9306229 PERCN6356473 PERCN9303419 PERCN9303449 PERCN9306360 PERCN9306362 PERCN9306364
Autosampler caps and septa
Sluitingen Sluitingen voor chromatografievials
These septa have a very broad chemical resistance and can be used in many markets including environmental, diagnostic packaging, pharmaceutical packaging, cosmetic and food packaging.
  • Provides excellent resealability
  • Multiple injection capability
  • Screw thread vial caps features Inter-Seal®
No adhesives are used in this process, bonding the cap and septa at the molecular level of plastic and rubber.