Connecting adapters, with parallel necks
Leverancier: NEUBERT VOLUME GLASSWAERE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0900-10-292929 0900-10-191929
NEUB0900-10-292929EA 111 EUR
NEUB0900-10-292929 NEUB0900-10-191929
Connecting adapters, with parallel necks
Adapters Adapters met NS-verbindingen
Two necked adapters made of borosilicate glass 3.3
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Order Now
Specificaties
Environmentally Preferable