Tubing reducer, T-shaped
Leverancier: PIEPER FILTER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PP-S16S24T PP-S16S32T PP-S24S16T PP-S24S32T PP-S32S16T PP-S32S24T PP-S16S24TL PP-S16S32TL PP-S24S32TL
229-4312EA 63.1 EUR
229-4312 229-4313 229-4321 229-4323 229-4331 229-4332 229-4341 229-4351 229-4352
Tubing reducer, T-shaped
Fittingen en connectoren
Made of PP, translucent.
  • Good temperature resistance
  • Good chemical resistance
T-shaped reducer for tubing.