Viscosity standards, VHG
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VHG-VISC20-500 VHG-VISC60A VHG-VISC900 VHG-VISC30-500 VHG-VISC120 VHG-VISC10-500 VHG-VISC50-500
PROOVHG-VISC20-500EA 0 EUR
PROOVHG-VISC20-500 PROOVHG-VISC60A PROOVHG-VISC900 PROOVHG-VISC30-500 PROOVHG-VISC120 PROOVHG-VISC10-500 PROOVHG-VISC50-500
Viscosity standards, VHG
Standaarden Standaarden voor viscometrie
These standard solutions are intended for use as a certified reference materiasl and calibration standards for viscometric instrumentation, using ASTM Method D445 and other viscometric methods.
Certificaten: Accredited to ISO 9001, ISO 17034 and ISO/IEC 17025. Traceable to NIST.