Bottle top dispensers, Dispensette® S, DAkkS
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Dispensette® S
BRNDDAKKS4600361EA 725 EUR
BRNDDAKKS4600361 BRNDDAKKS4600350 BRNDDAKKS4600351 BRNDDAKKS4600341 BRNDDAKKS4600331 BRNDDAKKS4600170 BRNDDAKKS4600171 BRNDDAKKS4600360
Bottle top dispensers, Dispensette® S, DAkkS
Flesdispensers
Fixed volume or variable volume, analogue-adjustable or Digital Easy Calibration.
  • With or without recirculation valve
  • Three different models of volume adjustment available: Fixed (Fixed-volume for standard applications; simple calibration adjustment with supplied tool) and Analog (Analog volume display, fast volume adjustment; simple calibration adjustment with supplied tool)
  • Digital EASY CALIBRATION: Easy to read digital display; dispensing volume can be set accurately and reproducibly. Easy Calibration technique for adjustment in seconds without tools; alteration of factory setting is automatically indicated
  • DE-M marking
  • Easy to calibrate and adjust in order to comply with ISO 9001 and GLP guidelines; alteration of factory setting is automatically indicated
Dispensette® S supports a very wide range of applications for the dispensing of aggressive reagents directly from the supply bottle: such as concentrated bases and acids like H₃PO₄, H₂SO₄ (with certain exceptions such as HCl, HNO₃, HF, etc.), saline solutions, and a variety of organic solvents. Parts that are in contact with the liquid are made from borosilicate glass, ceramic, platinum-iridium, ETFE, FEP, PFA, PTFE and PP (discharge tube safety screw cap). Colour code: Red.
Certificaten: Dispensette® S supplied with DE-M marking, performance certificate, telescopic filling tube, recirculation tube (optional), mounting tool and adapters of PP and DAkkS calibration certificate.