Clamp-on breaker lockout, EZ Panel Loc™
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
051254 065810
BRDY051254EA 30.46 EUR
BRDY051254 BRDY065810
Clamp-on breaker lockout, EZ Panel Loc™
Veiligheidssloten
Lockouts for use on breakers that do not have a hole in the switch tongue, including multipole breakers with tie-bars.
  • Turn thumbscrew to clamp lockout securely onto switch, then snap the cover closed to prevent thumbwheel from being loosened
  • Made from high-impact Isoplast Polymers