Handheld refractometers for wine, MASTER-P
Leverancier: Atago
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MASTER
ATAG2603EA 153 EUR
ATAG2603 ATAG2604 ATAG2601 ATAG2602 ATAG2600
Handheld refractometers for wine, MASTER-P
refractometers Draagbare refractometers
These refractometers are made of plastic and suitable for measuring grape must concentrations.
  • Dual scales: One wine scale - One Brix scale
  • Easy-to-read, smooth sampling and nice design
  • Improved hygiene aspect with a special grip
Simply apply a drop of sample on the prism to view the measurement value through the eyepiece.