Winkler electrodes, platinum/iridium
Leverancier: CHEMPUR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EI01/2 EI02
662-8421EA 0 EUR
662-8421 662-8422
Winkler electrodes, platinum/iridium
Elektroden Andere elektroden
This design consists of of a slit gauze cylinder containing a concentrically positioned wire spiral.
  • Electrode material platinum-iridium 90/10 ensures mechanical stability