PCR Master Mix, sparQ HiFi
Leverancier: Quantabio
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
95192-250 95192-050
733-2607EA 437 EUR
733-2607 733-2606
PCR Master Mix, sparQ HiFi
Reagentia voor nucleïnezuren Next generation reagentia voor sequencing
sparQ HiFi PCR Master Mix was specially designed for high efficiency library amplification from low DNA input. sparQ HiFi PCR Master Mix minimises the number of amplification cycles required to achieve the threshold required for sequencing. The result is a superior performing high fidelity master mix that increases library yield which reduces PCR-derived artifacts for a variety of sequencing applications.
  • Increased amplification efficiency results in higher yields for NGS library amplification, even from low input DNA
  • Unbiased amplification of DNA fragments provides improved coverage across AT- and GC-rich regions
  • Improved overall sequencing economics
The kit includes a primer mix allowing amplification of DNA libraries flanked by adapters containing the P5 and P7 Illumina® flow cell sequences.
Comparison of library preparation performed with sparQ HiFi PCR master mix matches PCR-free workflows. Lower bias enables better coverage uniformity resulting in greater sequencing depth or multiplexing capabilities.
Leveringsinformatie: Kit includes 2X HiFi PCR master mix, primer mix and HiFi enhancer.
Opgelet: sparQ HiFi PCR Master Mix is intended only for molecular biology applications and not for the diagnosis, prevention or treatment of a disease.