Multi-Elemental Standards, Metal Additives, CRM, VHG
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VHG-MA4-100G VHG-MA4-200G VHG-MA3-200G VHG-MA3-100G VHG-MA3-400G VHG-MA4-400G
PROOVHG-MA4-100GEA 0 EUR
PROOVHG-MA4-100G PROOVHG-MA4-200G PROOVHG-MA3-200G PROOVHG-MA3-100G PROOVHG-MA3-400G PROOVHG-MA4-400G
Multi-Elemental Standards, Metal Additives, CRM, VHG
Standaarden Kationische standaarden Standaarden met meerdere elementen
The solutions are intended for use as a certified reference material or calibration standard for the analysis of these elements in petroleum products or other organic matrices using inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), rotating disc electrode atomic emission spectroscopy (RDE-AES), flame or furnace atomic absorption spectroscopy (AA or GFAA), and other techniques for elemental analysis.
Certificaten: Traceable to NIST, SRM.