Empty racks for tipfill system, Qualitix®
Leverancier: Socorex Isba
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3070.0210ER 3070.1000ER
SOCO3070.0210EREA 47.6 EUR
SOCO3070.0210ER SOCO3070.1000ER
Empty racks for tipfill system, Qualitix®
Pipette Tip Boxes and Racks
These empty racks are designed to be used with Qualitix® tipfill packs.
  • Racks have smooth rounded edges and ergonomic snap latch that won’t catch gloves
  • Transparent lid enables instant view of content
  • Empty racks are re-usable and autoclavable