Bailers
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A296-1 A296-2 A296-4
SAMPA296-1EA 113 EUR
SAMPA296-1 SAMPA296-2 SAMPA296-4
Bailers
Monsternemers Monsternemers voor vloeistoffen
These bailers are made from high quality 316 stainless steel.
  • Heavy duty construction with satin finish
  • Fully welded handles giving a hygienic crevice free finish
Bestelinformatie: Available in three formats.