Reference materials, diesel fuel dilution Standards, VHG
Leverancier: LGC Standards PROMOCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VHGDSLFDBLK100
VHGDSLFD5P-100
PROOVHGDSLFDBLK100EA
0
EUR
PROOVHGDSLFDBLK100
PROOVHGDSLFD5P-100
Reference materials, diesel fuel dilution Standards, VHG
Standaarden
Standaarden voor de petroleumindustrie
These standards are intended for use in the determination of fuel (diesel fuel or gasoline) in used engine oils by GC, IR, fuel dilution meter (fuel sniffer), or flash point analyser. They are prepared gravimetrically from well-characterised fuels and hydrocarbon oils, and are verified by GC-FID in accordance with ASTM Method D3524 or D3525.
Certificaten: Accredited to ISO 9001, ISO 17034, and ISO/IEC 17025, traceabile to NIST, SRM.