CHO cell culture platform liquid media, HyClone™ HyCell™ TransFx-C
Leverancier: HyClone products (Cytiva)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SH30941.02
SH30941.03
SH30942.01
SH30941.02EA
102
EUR
SH30941.02
SH30941.03
SH30942.01
CHO cell culture platform liquid media, HyClone™ HyCell™ TransFx-C
Celcultuurmedia
HyCell™ TransFx-C Media is a hydrolysate-free and regulatory-friendly media which supports transient transfection, growth, and expression of a variety of recombinant proteins in CHO cells.
- Defined hydrolosate-free ADCF cell culture media formulation
- Supports the transient expression of a variety of recombinant proteins
- Same media for transient transfection and production
- Developed through our Metabolic Pathway Design process
- Component traceability
High-performing, animal-derived component-free (ADCF) cell culture media with sodium bicarbonate and without L-glutamine and poloxamer 188.
Certificaten: Manufactured in cGMP (21 CFR 820) compliant and ISO 9001 certified facilities.