Plant cell culture media, Hoagland's modified basal salt mixture, MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
092621820 092621822
ICNA092621820EA 60.3 EUR
ICNA092621820 ICNA092621822
Plant cell culture media, Hoagland's modified basal salt mixture, MP Biomedicals
Celcultuurmedia
Hoagland's modified basal salt mixture with macro- and micronutrients used to support to facilitate plant growth.
Order Now
Specificaties
Environmentally Preferable