Multi-element standards, common and transition elements, VHG
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VHG-SM75B-100 VHG-SM75B-500
PROOVHG-SM75B-100EA 0 EUR
PROOVHG-SM75B-100 PROOVHG-SM75B-500
Multi-element standards, common and transition elements, VHG
Standaarden Kationische standaarden Standaarden met meerdere elementen
This standard solution is intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), or alternative techniques, such as and flame or furnace atomic absorption spectroscopy (AA or GFAA).
Multi elements standards for Ag, Al, As, Ba, Be, Bi, Cd, Cr, Co, Cu, Fe, Li, Mn, Mo, Ni, Pb, Sb, Se, Sr, Tl, V, Zn at 100 µg/ml and Ca, K, Mg, Na at 1000 µg/ml in 0,2% HF.
Certificaten: Accredited to ISO 9001, ISO 17034 and ISO/IEC 17025. Traceable to NIST and SRM.