Multi-element standards, comprehensive mix B, VHG
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VHG-SM90C-500 VHG-SM90C-100
PROOVHG-SM90C-500EA 0 EUR
PROOVHG-SM90C-500 PROOVHG-SM90C-100
Multi-element standards, comprehensive mix B, VHG
Standaarden Kationische standaarden Standaarden met meerdere elementen
This standard solution is intended for use as a certified reference material and calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), or alternative techniques, such as and flame or furnace atomic absorption spectroscopy (AA or GFAA).
Multi element standards for Ag, Al, As, Ba, Bi, Ca, Cd, Ce, Dy, Er, Eu, Ga, Gd, Ho, La, Lu, Mg, Na, Nd, P, Pb, Pr, Rb, Sc, Se, Sm, Sr, Tb, Th, Tl, Tm, U, Y and Yb at 10 µg/ml in 40% aqua regia, tr. HF.
Certificaten: Accredited to ISO 9001, ISO 17034 and ISO/IEC 17025. Traceable to NIST and SRM.