Lubricants, APIEZON®
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C986.1 C987.1 1683.1 1686.1
ROTHC986.1EA 434 EUR
ROTHC986.1 ROTHC987.1 ROTH1683.1 ROTH1686.1
Lubricants, APIEZON®
Smeermiddelen
High lubricating and gettering properties. Extremely low steam pressures.
  • Chemically inert
  • Silicone-free
  • Non toxic
  • Temperature resistant
  • 10 years shelf life