Elemental analysis standards, soot content standards, VHG
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VHG-SOOT-SET VHG-SOOT-A-50 VHG-SOOT-B-50 VHG-SOOT-C-50 VHG-SOOT-D-50 VHG-SOOT-E-50
PROOVHG-SOOT-SETEA 0 EUR
PROOVHG-SOOT-SET PROOVHG-SOOT-A-50 PROOVHG-SOOT-B-50 PROOVHG-SOOT-C-50 PROOVHG-SOOT-D-50 PROOVHG-SOOT-E-50
Elemental analysis standards, soot content standards, VHG
Standaarden Standaarden voor de petroleumindustrie
This reference or calibration standard is intended for use in the determination of soot in diesel motor oil by infrared spectroscopy and other technique.
Certificaten: Conforms to ISO 9001 and ISO/IEC 17025.