All weather outdoor jacket, Sealtex™ Ultra, S496
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S496YNRXL S496YNRXXL S496YNRL
PRTWS496YNRXLEA 88.5 EUR
PRTWS496YNRXL PRTWS496YNRXXL PRTWS496YNRL
All weather outdoor jacket, Sealtex™ Ultra, S496
Werkkleding
This two-tone jacket made from lightweight Sealtex™ Ultra fabric (high technology PU coated fabric on a flexible 100% polyester Tricot knit) offers breathability as well as waterproof and windproof protection. The attached hood and studded storm flap are great features in bad weather.
  • Waterproof with welded seams to prevent water penetration
  • Heat-sealed reflective tape for added visibility
  • Two pockets for secure storage (deep storage pockets)
  • Two-way zip for quick and easy access; storm flap front to protect against the elements
  • Pack away hood for added functionality
  • Stud adjustable cuffs for a secure fit
  • Contrast panels for protection against dirt
Weight: 185 g/m²
Certificaten: EN 343 (3:2), EN ISO 20471 (Class 3)