Thallium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VHG-PTLN-100 VHG-PTLN-500 VHG-TTLN-100 VHG-TTLN-500
PROOVHG-PTLN-100EA 0 EUR
PROOVHG-PTLN-100 PROOVHG-PTLN-500 PROOVHG-TTLN-100 PROOVHG-TTLN-500
Thallium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Standaarden Kationische standaarden Thallium standaarden
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certificaten: Traceable to NIST, SRM.
Order Now
Specificaties
Environmentally Preferable