Tissue cool plate, COP 30
Leverancier: Medite
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
632-7014EA 4080 EUR
632-7014
Tissue cool plate, COP 30
Koelers Tafelmodel koelers
This tissue cool plate is designed for use for the rapid cooling of paraffin blocks during tissue processing or sectioning.
  • Quiet, efficient and environmentally-friendly
  • Can perform a cooling temperature down to –15 °C
  • Variable digital temperature control
Made of polished stainless steel with heated brims to prevent condensation, this plate enables the cooling of 60 to 100 embedding molds or paraffin blocks simultaneously.