Volumetric flasks, class A, glass, amber, NS glass stopper, BLAUBRAND®
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
937448 937443 937449 937453 937446 937450 937447 937451 937452 937445 937438
BRND937448EA 64.5 EUR
BRND937448 BRND937443 BRND937449 BRND937453 BRND937446 BRND937450 BRND937447 BRND937451 BRND937452 BRND937445 BRND937438
Volumetric flasks, class A, glass, amber, NS glass stopper, BLAUBRAND®
Kolven Maatkolven
Borosilicate glass 3.3, amber, with glass stopper.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in high contrast white enamel
  • DE-M marking
*Wide mouth.
Certificaten: DIN EN ISO 1042.
Bestelinformatie: Supplied with an individual certificate.