Silicon standard solution, for ICP, in H₂O, trace F⁻, CRM, VHG
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VHG-PSIW-100 VHG-PSIW-500 VHG-TSIW-100 VHG-TSIW-500
PROOVHG-PSIW-100EA 0 EUR
PROOVHG-PSIW-100 PROOVHG-PSIW-500 PROOVHG-TSIW-100 PROOVHG-TSIW-500
Silicon standard solution, for ICP, in H₂O, trace F⁻, CRM, VHG
Standaarden Kationische standaarden Silicium standaarden
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certificaten: Traceable to NIST, SRM.