Samarium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VHG-PSMN-100 VHG-PSMN-500 VHG-TSMN-100 VHG-TSMN-500
PROOVHG-PSMN-100EA 0 EUR
PROOVHG-PSMN-100 PROOVHG-PSMN-500 PROOVHG-TSMN-100 PROOVHG-TSMN-500
Samarium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Standaarden Kationische standaarden Samarium standaarden
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certificaten: Traceable to NIST, SRM.