Single element petroleum standards, silicon in hydrocarbon oil, CRM, VHG
Leverancier: LGC Standards PROMOCHEMTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
This solution is intended for use as a certified reference material or calibration standard for the analysis of this element in petroleum products or other organic matrices using inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), rotating disc electrode atomic emission spectroscopy (RDE-AES), flame or furnace atomic absorption spectroscopy (AA or GFAA), and other techniques for elemental analysis.
Certificaten: Traceable to NIST, SRM.