Petroleum standards, heavy mineral oils, CRM
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VHG-OIL-75-500 VHG-OIL-751GAL
PROOVHG-OIL-75-500EA 0 EUR
PROOVHG-OIL-75-500 PROOVHG-OIL-751GAL
Petroleum standards, heavy mineral oils, CRM
Standaarden Standaarden voor de petroleumindustrie
This matrix material solution is intended for use as a diluent for other metallo-organic standards.
Certificaten: ISO 9001 and ISO/IEC 17025 Certified and traceable to NIST.