Scintillation cocktail, Ultima Gold LLT
Leverancier: PerkinElmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6013371 6013377
PERK6013371EA 121 EUR
PERK6013371 PERK6013377
Scintillation cocktail, Ultima Gold LLT
Scintillatiecocktails
A biodegradable, high flash-point liquid scintillation cocktail which allows the user to determine low levels of ³H in a wide range of water samples without requiring distillation.
  • High water capacity - up to 54%
  • High counting efficiency
  • Very low background levels
  • For bioassay of urine up to 40% capacity, allowing detection levels of less than 10Bq/L-1 (500 minute count time)
  • Heavy metal radionuclide sample capacity, up to 18% of 2M HNO3, 2M HCl and 2M H3PO4 for alpha, beta and alpha/beta counting
  • No diffusion through polyethylene vials
  • High flash-point of approximately 140°C
Ultima Gold™ LLT accepts up to 54% tap water, river water, rain water and even sea water, with ³H counting efficiencies of approximately 30% and with very low background levels. When used with the Tri-Carb® time-resolved low-level liquid scintillation analyzers, backgrounds are reduced even further, resulting in minimum detectable activities of less than 1.1Bq/L-1 (500 minute count time).